missing translation for 'onlineSavingsMsg'
Learn More

L-Histidine, 98.5-101.5%, Spectrum™ Chemical

Catalog No. 18611283
Change view
Click to view available options
Quantity:
25 g
100 g
1 kg
5 kg
Packaging:
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18611283 25 g Poly Pail
18611282 100 g Poly Pail
18611281 1 kg Poly Pail
18611284 5 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18611283 Supplier Spectrum Chemical Mfg Cor Supplier No. HI12025GM
Get Up to $500 in Chemicals with Purchase
View all promotions

Chemical Identifiers

CAS 71-00-1

Specifications

CAS Max % 1
Heavy Metals 1.5e-05
Loss on Drying 0.002
Packaging Poly Pail
Specific Rotation +12.6° to +14.0°
Quantity 25 g
Percent Purity 98.5 to 101.5%
Grade Reagent
Chemical Name or Material L-Histidine

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.