missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-(-)-Fucose 97.0+%, TCI America™
Supplier: TCI America F006510G
| Quantity | 10 g |
|---|
Chemical Identifiers
| C6H12O5 | |
| MFCD00135607 | |
| l---fucose, aldehydo-l-fucose, l-galactose, 6-deoxy, 2s,3r,4r,5s-2,3,4,5-tetrahydroxyhexanal, unii-28ryy2iv3f, l---rhodeose, 28ryy2iv3f, l--fucose, l-rhodeose, 3h-fucose | |
| CHEBI:48204 | |
| C[C@@H]1OC(O)[C@@H](O)[C@H](O)[C@@H]1O |
Specifications
| L-(-)-Fucose | |
| 140°C | |
| C6H12O5 | |
| MFCD00135607 | |
| SHZGCJCMOBCMKK-DHVFOXMCSA-N | |
| (3S,4R,5S,6S)-6-methyloxane-2,3,4,5-tetrol | |
| 3034656 | |
| 164.16 | |
| Crystalline Powder |
| 2438-80-4 | |
| White | |
| 10 g | |
| l---fucose, aldehydo-l-fucose, l-galactose, 6-deoxy, 2s,3r,4r,5s-2,3,4,5-tetrahydroxyhexanal, unii-28ryy2iv3f, l---rhodeose, 28ryy2iv3f, l--fucose, l-rhodeose, 3h-fucose | |
| C[C@@H]1OC(O)[C@@H](O)[C@H](O)[C@@H]1O | |
| 164.16 | |
| CHEBI:48204 | |
| ≥97.0% (HPLC) |
Safety and Handling
TSCA : Yes