missing translation for 'onlineSavingsMsg'
Learn More

L-(-)-Fucose 97.0+%, TCI America™

Catalog No. p-7129473
Change view
Click to view available options
Quantity:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F00651G 1 g
F006510G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F00651G Supplier TCI America Supplier No. F00651G

Chemical Identifiers

CAS 2438-80-4
Molecular Formula C6H12O5
Molecular Weight (g/mol) 164.16
MDL Number MFCD00135607
InChI Key SHZGCJCMOBCMKK-DHVFOXMCSA-N
Synonym l---fucose, aldehydo-l-fucose, l-galactose, 6-deoxy, 2s,3r,4r,5s-2,3,4,5-tetrahydroxyhexanal, unii-28ryy2iv3f, l---rhodeose, 28ryy2iv3f, l--fucose, l-rhodeose, 3h-fucose
PubChem CID 3034656
ChEBI CHEBI:48204
IUPAC Name (3S,4R,5S,6S)-6-methyloxane-2,3,4,5-tetrol
SMILES C[C@@H]1OC(O)[C@@H](O)[C@H](O)[C@@H]1O

Specifications

Melting Point 140°C
Color White
Quantity 1 g
Formula Weight 164.16
Percent Purity ≥97.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material L-(-)-Fucose