missing translation for 'onlineSavingsMsg'
Learn More

L(-)-Epinephrine, 99%

Catalog No. p-125355
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC204400010 1 g Glass bottle
AC204400050 5 g Glass bottle
AC204400250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC204400010 Supplier Thermo Scientific Chemicals Supplier No. 204400010
Only null left

CAS: 51-43-4 | C9H13NO3 | 183.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 51-43-4
MDL Number MFCD00002204
InChI Key UCTWMZQNUQWSLP-VIFPVBQESA-N
Synonym epinephrine, l-adrenaline, adrenaline, l-epinephrine, adrenalin, levoepinephrine, epinefrina, --adrenaline, adnephrine, chelafrin
PubChem CID 5816
ChEBI CHEBI:28918
IUPAC Name 4-[(1R)-1-hydroxy-2-(methylamino)ethyl]benzene-1,2-diol
SMILES CNCC(C1=CC(=C(C=C1)O)O)O

Specifications

Melting Point 208.0°C to 211.0°C
Color White to Beige
Loss on Drying 1% max. (18 hrs) (0.7 kPa, P2O5)
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Molecular Formula C9H13NO3
Linear Formula (HO)2C6H3CH(CH2NHCH3)OH
Quantity 1 g
Beilstein 13, 830
Merck Index 15, 3674
Specific Rotation Condition − 51.50 (20.00°C c=4,1M HCl,dry sub)
Specific Rotation − 51.50
Solubility Information Solubility in water: sparingly soluble. Other solubilities: insoluble in alcohol, chloroform, ether and, acetone
Molecular Weight (g/mol) 183.21
Formula Weight 183.21
Percent Purity 99%
Physical Form Fine Crystalline Powder
Chemical Name or Material L(-)-Epinephrine, 99%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes skin irritation. Toxic if inhaled. Toxic if swallowed. Toxic in contact with skin. Causes serious eye irritation. May cause respiratory irritation. Harmful to aquatic life with long lasting effects. May cause damage to organs through prolonged or repeated exposure.
Precautionary Statement GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician. Wear protective gloves/protective clothing/eye protection/face protection. Call a POISON CENTER or doctor/physician if you feel unwell. IF ON SKIN: Gently wash with plenty of soap and water. IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing. Avoid breathing dust/fume/gas/mist/vapors/spray. Do not breathe dust/fume/gas/mist/vapors/spray. Avoid release to the environment. IF ON SKIN: Wash with plenty of soap and water. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
EINECSNumber 200-098-7
RTECSNumber DO2625000
TSCA TSCA
Recommended Storage product darkens on exposure to air and light

RUO – Research Use Only