accessibility menu, dialog, popup

UserName

CAS RN 51-43-4

IUPAC Name: 4-[(1R)-1-hydroxy-2-(methylamino)ethyl]benzene-1,2-diol
Synonyms: epinephrine racepinefrine soladren bernarenin adrine (R)-(-)-adnephrine D-adrenaline norartrinal supranefran epinephrine preparation (product)
Molecular Weight (g/mol): 183.21
Molecular Formula: C9H13NO3
InChi Key: UCTWMZQNUQWSLP-VIFPVBQESA-N
SMILES: CNC[C@H](O)C1=CC=C(O)C(O)=C1
Molecular Weight (g/mol) 
  • (2)
  • (3)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (5)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (3)