missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ L-DOPA
Description
Immediate precursor of dopamine, produced by tyrosine hydroxylase. Displays antiParkinsonian activity.
Specifications
Specifications
| Quantity | 50 mg |
| Product Line | Tocris |
| Formula Weight | 197.19 |
| Purity | >99% |
| Molecular Weight (g/mol) | 197.19 |
| Chemical Name or Material | L-DOPA |
Safety and Handling
| Recommended Storage | Store at -20°C |