missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Citrulline 98.0+%, TCI America™
Supplier: TCI America C0372250G
Specifications
| L-Citrulline | |
| White | |
| MFCD00064397 | |
| l-citrulline, citrulline, h-cit-oh, delta-ureidonorvaline, sitrulline, n5-carbamoyl-l-ornithine, l-cytrulline, n5-aminocarbonyl ornithine, n delta-carbamylornithine, citrulline, l | |
| NC(CCCNC(N)=O)C(O)=O | |
| 175.19 | |
| CHEBI:16349 | |
| ≥98.0% (T) |
| 372-75-8 | |
| C6H13N3O3 | |
| 250 g | |
| RHGKLRLOHDJJDR-UHFFFAOYNA-N | |
| 2-amino-5-(carbamoylamino)pentanoic acid | |
| 9750 | |
| 175.19 | |
| Crystalline Powder |
Chemical Identifiers
| 372-75-8 | |
| 175.19 | |
| RHGKLRLOHDJJDR-UHFFFAOYNA-N | |
| 9750 | |
| 2-amino-5-(carbamoylamino)pentanoic acid |
| C6H13N3O3 | |
| MFCD00064397 | |
| l-citrulline, citrulline, h-cit-oh, delta-ureidonorvaline, sitrulline, n5-carbamoyl-l-ornithine, l-cytrulline, n5-aminocarbonyl ornithine, n delta-carbamylornithine, citrulline, l | |
| CHEBI:16349 | |
| NC(CCCNC(N)=O)C(O)=O |
Safety and Handling
EINECSNumber : (9)-0503
TSCA : Yes