missing translation for 'onlineSavingsMsg'
Learn More

L-Citrulline 98.0+%, TCI America™

Catalog No. p-7124380
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C037225G 25 g
C0372250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C037225G Supplier TCI America Supplier No. C037225G
Only null left

Chemical Identifiers

CAS 372-75-8
Molecular Formula C6H13N3O3
Molecular Weight (g/mol) 175.19
MDL Number MFCD00064397
InChI Key RHGKLRLOHDJJDR-UHFFFAOYNA-N
Synonym l-citrulline, citrulline, h-cit-oh, delta-ureidonorvaline, sitrulline, n5-carbamoyl-l-ornithine, l-cytrulline, n5-aminocarbonyl ornithine, n delta-carbamylornithine, citrulline, l
PubChem CID 9750
ChEBI CHEBI:16349
IUPAC Name 2-amino-5-(carbamoylamino)pentanoic acid
SMILES NC(CCCNC(N)=O)C(O)=O

Specifications

Color White
Quantity 25 g
Formula Weight 175.19
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material L-Citrulline
EINECSNumber (9)-0503
TSCA Yes