missing translation for 'onlineSavingsMsg'
Learn More

L-Aspartic acid β-benzyl ester, 98%, Thermo Scientific™

Chemical Identifiers

CAS 2177-63-1
Molecular Formula C11H13NO4
Molecular Weight (g/mol) 223.23
InChI Key VGALFAWDSNRXJK-VIFPVBQESA-N
Synonym h-asp obzl-oh, l-aspartic acid 4-benzyl ester, l-aspartic acid beta-benzyl ester, beta-benzyl l-aspartate, 2s-2-amino-4-benzyloxy-4-oxobutanoic acid, 4-benzyl l-aspartate, benzyl hydrogen beta-l-aspartate, s-2-amino-4-benzyloxy-4-oxobutanoic acid, 2s-2-amino-4-oxo-4-phenylmethoxybutanoic acid, asp obzl
PubChem CID 101186
IUPAC Name (2S)-2-amino-4-oxo-4-phenylmethoxybutanoic acid
SMILES C1=CC=C(C=C1)COC(=O)CC(C(=O)O)N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC441930050
SDS View Documents
Thermo Scientific Chemicals
441930050
5g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC441930250
SDS View Documents
Thermo Scientific Chemicals
441930250
25g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

2177-63-1
223.23
h-asp obzl-oh, l-aspartic acid 4-benzyl ester, l-aspartic acid beta-benzyl ester, beta-benzyl l-aspartate, 2s-2-amino-4-benzyloxy-4-oxobutanoic acid, 4-benzyl l-aspartate, benzyl hydrogen beta-l-aspartate, s-2-amino-4-benzyloxy-4-oxobutanoic acid, 2s-2-amino-4-oxo-4-phenylmethoxybutanoic acid, asp obzl
(2S)-2-amino-4-oxo-4-phenylmethoxybutanoic acid
C11H13NO4
VGALFAWDSNRXJK-VIFPVBQESA-N
101186
C1=CC=C(C=C1)COC(=O)CC(C(=O)O)N

Specifications

2177-63-1
97.5% min. (HPLC)
5g
h-asp obzl-oh, l-aspartic acid 4-benzyl ester, l-aspartic acid beta-benzyl ester, beta-benzyl l-aspartate, 2s-2-amino-4-benzyloxy-4-oxobutanoic acid, 4-benzyl l-aspartate, benzyl hydrogen beta-l-aspartate, s-2-amino-4-benzyloxy-4-oxobutanoic acid, 2s-2-amino-4-oxo-4-phenylmethoxybutanoic acid, asp obzl
C1=CC=C(C=C1)COC(=O)CC(C(=O)O)N
223.23
223.23
L-Aspartic acid β-benzyl ester
Authentic
C11H13NO4
+ 27.8
VGALFAWDSNRXJK-VIFPVBQESA-N
(2S)-2-amino-4-oxo-4-phenylmethoxybutanoic acid
101186
98%
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 218-541-8

SDS
Documents

Documents

RUO – Research Use Only