accessibility menu, dialog, popup

UserName

CAS RN 2177-63-1

IUPAC Name: 2-amino-4-(benzyloxy)-4-oxobutanoic acid
Synonyms: 2-amino-4-(benzyloxy)-4-oxobutanoic acid
Molecular Weight (g/mol): 223.23
Molecular Formula: C11H13NO4
InChi Key: VGALFAWDSNRXJK-UHFFFAOYNA-N
SMILES: NC(CC(=O)OCC1=CC=CC=C1)C(O)=O
Molecular Weight (g/mol) 
  • (4)
  • (2)
Percent Purity 
  • (2)
  • (4)
Quantity 
  • (3)
  • (3)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (4)
  • (4)
  • (2)
  • (2)
  • (4)
  • (3)
  • (3)
  • (2)
  • (2)