missing translation for 'onlineSavingsMsg'
Learn More

L-Ascorbic Acid ACS AR, Macron Fine Chemicals™

macron

Chemical Identifiers

CAS 50-81-7
Molecular Formula C6H8O6
Molecular Weight (g/mol) 176.124
InChI Key CIWBSHSKHKDKBQ-JLAZNSOCSA-N
Synonym l-ascorbic acid, ascorbic acid, vitamin c, l-ascorbate, ascorbate, ascorbicap, l +-ascorbic acid, cevitamic acid, ascoltin, hybrin
PubChem CID 54670067
ChEBI CHEBI:29073
IUPAC Name (2R)-2-[(1S)-1,2-dihydroxyethyl]-3,4-dihydroxy-2H-furan-5-one
SMILES C(C(C1C(=C(C(=O)O1)O)O)O)O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
MK4407100
View Documents
Avantor Macron
440702
100g Glass N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

50-81-7
176.124
l-ascorbic acid, ascorbic acid, vitamin c, l-ascorbate, ascorbate, ascorbicap, l +-ascorbic acid, cevitamic acid, ascoltin, hybrin
CHEBI:29073
C(C(C1C(=C(C(=O)O1)O)O)O)O
C6H8O6
CIWBSHSKHKDKBQ-JLAZNSOCSA-N
54670067
(2R)-2-[(1S)-1,2-dihydroxyethyl]-3,4-dihydroxy-2H-furan-5-one

Specifications

50-81-7
l-ascorbic acid, ascorbic acid, vitamin c, l-ascorbate, ascorbate, ascorbicap, l +-ascorbic acid, cevitamic acid, ascoltin, hybrin
C(C(C1C(=C(C(=O)O1)O)O)O)O
176.124
CHEBI:29073
ACS
C6H8O6
CIWBSHSKHKDKBQ-JLAZNSOCSA-N
(2R)-2-[(1S)-1,2-dihydroxyethyl]-3,4-dihydroxy-2H-furan-5-one
54670067
176.13
L-Ascorbic Acid
Videos
SDS
Documents

Documents