missing translation for 'onlineSavingsMsg'
Learn More

L-Arginine Hydrochloride, 98.5-101.5%, Spectrum™ Chemical

$194.89 - $1,007.64

Chemical Identifiers

CAS 1119-34-2
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
18611201
View Documents Promotion Details
N/A 25 g Poly Pail
Each for $194.89
Only null left
 
18611200
View Documents Promotion Details
N/A 100 g Poly Pail
Each for $359.61
Only null left
 
18611199
View Documents Promotion Details
N/A 1 kg Poly Pail
Each for $1,007.64
Only null left
 
Description

Description

Specifications

Chemical Identifiers

1119-34-2

Specifications

1119-34-2
20 ppm
+21.3° to +23.5°
Reagent
1
0.003
98.5 to 101.5%
L-Arginine Hydrochloride
Videos
SDS
Documents

Documents

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.