missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Arginine Hydrochloride, 98.5-101.5%, Spectrum™ Chemical
Supplier: Spectrum Chemical Mfg Cor AR1251KG
| Quantity | 1 kg |
|---|---|
| Packaging | Poly Pail |
Chemical Identifiers
| 1119-34-2 |
Specifications
| L-Arginine Hydrochloride | |
| 1 | |
| 0.003 | |
| +21.3° to +23.5° | |
| 98.5 to 101.5% |
| 1119-34-2 | |
| 20 ppm | |
| Poly Pail | |
| 1 kg | |
| Reagent |