missing translation for 'onlineSavingsMsg'
Learn More

L-Arginine, 98.5-101.5%, Spectrum™ Chemical

$198.75 - $27,541.07

Chemical Identifiers

CAS 74-79-3
View More Specs

Products 5
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
18611197
View Documents Promotion Details
N/A 25 g Amber Glass Bottle
Each for $198.75
Only null left
 
18611195
View Documents Promotion Details
N/A 100 g Amber Glass Bottle
Each for $374.50
Only null left
 
18611194
View Documents Promotion Details
N/A 1 kg Amber Glass Bottle
Each for $1,202.46
Only null left
 
18611196
View Documents Promotion Details
N/A 12 kg Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
18611198
View Documents Promotion Details
N/A 50 kg Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

74-79-3

Specifications

74-79-3
20 ppm
0.01
+26.0° to +27.7°
Reagent
1
10 ppm
0.002
98.5 to 101.5%
L-Arginine
Videos
SDS
Documents

Documents

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.