missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Arginine, 98.5-101.5%, Spectrum™ Chemical
Supplier: Spectrum Chemical Mfg Cor AR1201KG
| Quantity | 1 kg |
|---|---|
| Packaging | Amber Glass Bottle |
Chemical Identifiers
| 74-79-3 |
Specifications
| L-Arginine | |
| 1 | |
| 10 ppm | |
| Amber Glass Bottle | |
| +26.0° to +27.7° | |
| 98.5 to 101.5% |
| 74-79-3 | |
| 20 ppm | |
| 0.01 | |
| 0.002 | |
| 1 kg | |
| Reagent |