missing translation for 'onlineSavingsMsg'
Learn More

L-Arginine, 98.5-101.5%, Spectrum™ Chemical

Catalog No. 18611197
Change view
Click to view available options
Quantity:
25 g
100 g
1 kg
12 kg
50 kg
Packaging:
Amber Glass Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18611197 25 g Amber Glass Bottle
18611195 100 g Amber Glass Bottle
18611194 1 kg Amber Glass Bottle
18611196 12 kg Amber Glass Bottle
18611198 50 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18611197 Supplier Spectrum Chemical Mfg Cor Supplier No. AR12025GM

Chemical Identifiers

CAS 74-79-3

Specifications

CAS Max % 1
Heavy Metals 20 ppm
Lead (Pb) 10 ppm
Loss on Drying 0.01
Packaging Amber Glass Bottle
Residue after Ignition 0.002
Specific Rotation +26.0° to +27.7°
Quantity 25 g
Percent Purity 98.5 to 101.5%
Grade Reagent
Chemical Name or Material L-Arginine
Show More Show Less

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.