missing translation for 'onlineSavingsMsg'
Learn More

L-(-)-Arabitol 97.0+%, TCI America™

$94.93 - $683.51

Chemical Identifiers

CAS 7643-75-6
Molecular Formula C5H12O5
Molecular Weight (g/mol) 152.15
MDL Number MFCD00064290
InChI Key HEBKCHPVOIAQTA-UHFFFAOYNA-N
Synonym l-arabitol, l-arabinitol, l-arabinol, l-lyxitol, l---arabitol, arabinitol, l---arabinitol, arabinitol, l, 2s,4s-pentane-1,2,3,4,5-pentol, l--arabitol
PubChem CID 439255
ChEBI CHEBI:18403
IUPAC Name pentane-1,2,3,4,5-pentol
SMILES OCC(O)C(O)C(O)CO
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
A05181G
View Documents Promotion Details
N/A 1 g
Each for $94.93
Only null left
 
A05185G
View Documents Promotion Details
N/A 5 g
Each for $214.54
Only null left
 
A051825G
View Documents Promotion Details
N/A 25 g
Each for $683.51
Only null left
 
Description

Description

Specifications

Chemical Identifiers

7643-75-6
152.15
HEBKCHPVOIAQTA-UHFFFAOYNA-N
439255
pentane-1,2,3,4,5-pentol
C5H12O5
MFCD00064290
l-arabitol, l-arabinitol, l-arabinol, l-lyxitol, l---arabitol, arabinitol, l---arabinitol, arabinitol, l, 2s,4s-pentane-1,2,3,4,5-pentol, l--arabitol
CHEBI:18403
OCC(O)C(O)C(O)CO

Specifications

7643-75-6
White
MFCD00064290
HEBKCHPVOIAQTA-UHFFFAOYNA-N
pentane-1,2,3,4,5-pentol
439255
152.15
Crystalline Powder
103°C
C5H12O5
l-arabitol, l-arabinitol, l-arabinol, l-lyxitol, l---arabitol, arabinitol, l---arabinitol, arabinitol, l, 2s,4s-pentane-1,2,3,4,5-pentol, l--arabitol
OCC(O)C(O)C(O)CO
152.15
CHEBI:18403
≥97.0% (GC)
L-(-)-Arabitol
Videos
Safety and Handling

Safety and Handling

TSCA : Yes

SDS
Documents

Documents