missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-(-)-Arabitol 97.0+%, TCI America™
Specifications
Specifications
| Melting Point | 103°C |
| Color | White |
| Quantity | 5 g |
| Formula Weight | 152.15 |
| Percent Purity | ≥97.0% (GC) |
| Grade | Not Available |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | L-(-)-Arabitol |
Safety and Handling
| TSCA | Yes |