missing translation for 'onlineSavingsMsg'
Learn More

L-(-)-Arabitol 97.0+%, TCI America™

Supplier:  TCI America A051825G

 View more versions of this product

Catalog No. A051825G


Only null left

Description

Description

Specifications

Specifications

L-(-)-Arabitol
103°C
C5H12O5
25 g
HEBKCHPVOIAQTA-UHFFFAOYNA-N
pentane-1,2,3,4,5-pentol
439255
152.15
Crystalline Powder
7643-75-6
White
MFCD00064290
l-arabitol, l-arabinitol, l-arabinol, l-lyxitol, l---arabitol, arabinitol, l---arabinitol, arabinitol, l, 2s,4s-pentane-1,2,3,4,5-pentol, l--arabitol
OCC(O)C(O)C(O)CO
152.15
CHEBI:18403
≥97.0% (GC)

Chemical Identifiers

7643-75-6
152.15
HEBKCHPVOIAQTA-UHFFFAOYNA-N
439255
pentane-1,2,3,4,5-pentol
C5H12O5
MFCD00064290
l-arabitol, l-arabinitol, l-arabinol, l-lyxitol, l---arabitol, arabinitol, l---arabinitol, arabinitol, l, 2s,4s-pentane-1,2,3,4,5-pentol, l--arabitol
CHEBI:18403
OCC(O)C(O)C(O)CO
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions