missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Alanine, FCC, 98.5-101.5%, Spectrum™ Chemical
A1252, 56-41-7, C3H7NO2
$232.60 - $4,743.42
Chemical Identifiers
| CAS | 56-41-7 |
|---|---|
| Molecular Formula | C3H7NO2 |
| Molecular Weight (g/mol) | 89.09 |
| MDL Number | MFCD00064410 |
| InChI Key | QNAYBMKLOCPYGJ-REOHCLBHSA-N |
| IUPAC Name | (2S)-2-aminopropanoic acid |
| SMILES | C[C@H](N)C(O)=O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|
18603702
|
N/A | 25 g | Amber Glass Bottle |
Each for $232.60
|
|
||||
|
18603703
|
N/A | 100 g | Poly Bottle |
Each for $367.56
|
|
||||
|
18603704
|
N/A | 1 kg | Poly Pail |
Each for $1,367.02
|
|
||||
|
18603705
|
N/A | 5 kg | Poly Pail |
Each for $4,743.42
|
|
||||
Description
Spectrum™ Chemical L-Alanine, FCC is included in the 20 amino acids encoded by the genetic code and often used in cases of hypoglycemia. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.Chemical Identifiers
| 56-41-7 | |
| 89.09 | |
| QNAYBMKLOCPYGJ-REOHCLBHSA-N | |
| C[C@H](N)C(O)=O |
| C3H7NO2 | |
| MFCD00064410 | |
| (2S)-2-aminopropanoic acid |
Specifications
| 56-41-7 | |
| 0.2% | |
| 0.3% | |
| MFCD00064410 | |
| QNAYBMKLOCPYGJ-REOHCLBHSA-N | |
| (2S)-2-aminopropanoic acid | |
| 98.5 to 101.5% |
| 100% | |
| 5 mg/kg | |
| C3H7NO2 | |
| +13.5° to +15.5° | |
| C[C@H](N)C(O)=O | |
| 89.09 | |
| FCC |