missing translation for 'onlineSavingsMsg'
Learn More

L-Alanine, FCC, 98.5-101.5%, Spectrum™ Chemical

Catalog No. 18603702
Change view
Click to view available options
Quantity:
25 g
100 g
1 kg
5 kg
Packaging:
Amber Glass Bottle
Poly Bottle
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18603702 25 g Amber Glass Bottle
18603703 100 g Poly Bottle
18603704 1 kg Poly Pail
18603705 5 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18603702 Supplier Spectrum Chemical Mfg Cor Supplier No. A125225GM
Get Up to $500 in Chemicals with Purchase
View all promotions

A1252, 56-41-7, C3H7NO2

Spectrum™ Chemical L-Alanine, FCC is included in the 20 amino acids encoded by the genetic code and often used in cases of hypoglycemia. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 56-41-7
Molecular Formula C3H7NO2
Molecular Weight (g/mol) 89.09
MDL Number MFCD00064410
InChI Key QNAYBMKLOCPYGJ-REOHCLBHSA-N
IUPAC Name (2S)-2-aminopropanoic acid
SMILES C[C@H](N)C(O)=O

Specifications

CAS Max % 100%
Ignition Residue 0.2%
Lead (Pb) 5 mg/kg
Loss on Drying 0.3%
Packaging Amber Glass Bottle
Specific Rotation +13.5° to +15.5°
Quantity 25 g
Percent Purity 98.5 to 101.5%
Grade FCC