missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals L-Alanine, 99%

CAS: 56-41-7 | C3H7NO2 | 89.09 g/mol

Supplier:  Thermo Scientific Chemicals A1580422

 View more versions of this product

Catalog No. AAA1580422


Only null left

Description

Description

L-Alanine plays a vital role in glucose-alanine cycle between tissue and liver. It is used for protein construction as well as involved in the metabolism of tryptophan and vitamin pyridoxine. It provides energy for muscles and central nervous system in order to strengthen the immune system. It helps in the metabolism of sugars and organic acids, and enhances the germination rates of Bacillus subtilis spores. It also displays a cholesterol-reducing effect in animals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-Alanine plays a vital role in glucose-alanine cycle between tissue and liver. It is used for protein construction as well as involved in the metabolism of tryptophan and vitamin pyridoxine. It provides energy for muscles and central nervous system in order to strengthen the immune system. It helps in the metabolism of sugars and organic acids, and enhances the germination rates of Bacillus subtilis spores. It also displays a cholesterol-reducing effect in animals.

Solubility
Soluble in water and cold ethanol. Slightly soluble in pyridine. Insoluble in ether and acetone.

Notes
Incompatible with strong oxidizing agents.
Specifications

Specifications

∼295°C (decomposition)
56-41-7
C3H7NO2
1720248
l-alanine, alanine, l-alpha-alanine, s-alanine, s-2-aminopropanoic acid, l-2-aminopropionic acid, 2s-2-aminopropanoic acid, h-ala-oh, l-+-alanine, l-2-aminopropanoic acid
QNAYBMKLOCPYGJ-REOHCLBHSA-N
C[C@H](N)C(O)=O
89.09
5950
+14° (c=5 in 5N HCl)
99%
L-Alanine
99%
MFCD00064410
14,204
Soluble in water and cold ethanol. Slightly soluble in pyridine. Insoluble in ether and acetone.
1.432
(2S)-2-aminopropanoic acid
100 g
CHEBI:16977
89.09

Chemical Identifiers

56-41-7
89.09
QNAYBMKLOCPYGJ-REOHCLBHSA-N
5950
(2S)-2-aminopropanoic acid
C3H7NO2
MFCD00064410
l-alanine, alanine, l-alpha-alanine, s-alanine, s-2-aminopropanoic acid, l-2-aminopropionic acid, 2s-2-aminopropanoic acid, h-ala-oh, l-+-alanine, l-2-aminopropanoic acid
CHEBI:16977
C[C@H](N)C(O)=O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 200-273-8

RTECSNumber : AY2990000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only