Learn More
Thermo Scientific Chemicals L-Alanine, 99%
Description
L-Alanine plays a vital role in glucose-alanine cycle between tissue and liver. It is used for protein construction as well as involved in the metabolism of tryptophan and vitamin pyridoxine. It provides energy for muscles and central nervous system in order to strengthen the immune system. It helps in the metabolism of sugars and organic acids, and enhances the germination rates of Bacillus subtilis spores. It also displays a cholesterol-reducing effect in animals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Soluble in water and cold ethanol. Slightly soluble in pyridine. Insoluble in ether and acetone.
Notes
Incompatible with strong oxidizing agents.
Specifications
Specifications
| Melting Point | ∽295°C (decomposition) |
| Assay Percent Range | 99% |
| Beilstein | 1720248 |
| Merck Index | 14,204 |
| Solubility Information | Soluble in water and cold ethanol. Slightly soluble in pyridine. Insoluble in ether and acetone. |
| Density | 1.432 |
| Quantity | 25 g |
| Formula Weight | 89.09 |
| Percent Purity | 99% |
| Chemical Name or Material | L-Alanine |
Safety and Handling
| EINECSNumber | 200-273-8 |
| RTECSNumber | AY2990000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only