missing translation for 'onlineSavingsMsg'
Learn More

L-Adrenaline, 98+%

Catalog No. AAL0491106
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0491106 5 g
AAL0491114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0491106 Supplier Thermo Scientific Chemicals Supplier No. L0491106

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 51-43-4 | C9H13NO3 | 183.207 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 51-43-4
Molecular Formula C9H13NO3
Molecular Weight (g/mol) 183.207
MDL Number MFCD00002204
InChI Key UCTWMZQNUQWSLP-VIFPVBQESA-N
Synonym epinephrine, l-adrenaline, adrenaline, l-epinephrine, adrenalin, levoepinephrine, epinefrina, --adrenaline, adnephrine, chelafrin
PubChem CID 5816
ChEBI CHEBI:28918
IUPAC Name 4-[(1R)-1-hydroxy-2-(methylamino)ethyl]benzene-1,2-diol
SMILES CNCC(C1=CC(=C(C=C1)O)O)O

Specifications

Melting Point 216°C to 219°C
Quantity 5 g
UN Number UN2811
Beilstein 2368277
Sensitivity Air and light sensitive
Merck Index 14,3619
Optical Rotation −52° (c=1 in 0.6N HCl)
Formula Weight 183.2
Percent Purity ≥98%
Chemical Name or Material L-Adrenaline
Hazard Category H301+H331-H310-H315-H319-H335-H373
Hazard Statement GHS H Statement
H300-H310-H330
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Precautionary Statement P260-P262-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P310-P311-P314-P330-P361-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 200-098-7
RTECSNumber DO2625000
TSCA Yes
Recommended Storage Keep cold; Store under Argon

RUO – Research Use Only