missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ L-838,417

Catalog No. 325010
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
325010 10 mg
325050 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 325010 Supplier Tocris Bioscience™ Supplier No. 3250/10
Only null left

GABAA partial agonist; displays subtype selectivity

Subtype-selective GABAA receptor partial agonist. Selectively binds to α1, α2, α3 and α5 subunits (Ki values are 0.79, 0.67, 0.67 and 2.25 nM respectively) but displays no efficacy at α1 (α1-sparing). Exhibits non-sedative anxiolytic, antinociceptive and anti-inflammatory activity in vivo.

Chemical Identifiers

CAS 286456-42-6
Molecular Formula C19H19F2N7O

Specifications

Quantity 10 mg
Formula Weight 399.4
Purity >98%
Molecular Weight (g/mol) 399.41
Chemical Name or Material L-838, 417
Recommended Storage Store at +4°C