missing translation for 'onlineSavingsMsg'
Learn More

L-(+)-2-Aminobutyric acid, 98+%

CAS: 1492-24-6 | C4H9NO2 | 103.121 g/mol

Supplier:  Thermo Scientific Chemicals L1398406

 View more versions of this product

Catalog No. AAL1398406


Only null left
Explore available promotions

Description

Description

L-(+)-2-Aminobutyric acid is used in the biosynthesis of nonribosomal peptides. It acts as a receptor antagonist. It is also used as a chiral reagent. Further, it is used in the determination of substrate of glutamyl cysteine acid synthase. In addition to this, it is also utilized as a drug intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-(+)-2-Aminobutyric acid is used in the biosynthesis of nonribosomal peptides. It acts as a receptor antagonist. It is also used as a chiral reagent. Further, it is used in the determination of substrate of glutamyl cysteine acid synthase. In addition to this, it is also utilized as a drug intermediate.

Solubility
Soluble in water and acetic acid. Slightly soluble in alcohol and ether.

Notes
Incompatible with strong oxidizing agents.
Specifications

Specifications

L-(+)-2-Aminobutyric acid
>300°C
MFCD00064415
1720935
l-2-aminobutyric acid, s-2-aminobutanoic acid, 2s-2-aminobutanoic acid, h-abu-oh, l-alpha-aminobutyric acid, s-+-2-aminobutyric acid, l-+-2-aminobutyric acid, --2-aminobutyric acid, s-2-aminobutyric acid, s-2-amino-butyric acid
QWCKQJZIFLGMSD-VKHMYHEASA-N
CCC(C(=O)O)N
103.121
CHEBI:35619
≥98%
1492-24-6
C4H9NO2
5 g
14,428
Soluble in water and acetic acid. Slightly soluble in alcohol and ether.
+21° (c=4.8 in 6N HCl)
(2S)-2-aminobutanoic acid
80283
103.12

Chemical Identifiers

1492-24-6
103.121
QWCKQJZIFLGMSD-VKHMYHEASA-N
80283
(2S)-2-aminobutanoic acid
C4H9NO2
MFCD00064415
l-2-aminobutyric acid, s-2-aminobutanoic acid, 2s-2-aminobutanoic acid, h-abu-oh, l-alpha-aminobutyric acid, s-+-2-aminobutyric acid, l-+-2-aminobutyric acid, --2-aminobutyric acid, s-2-aminobutyric acid, s-2-amino-butyric acid
CHEBI:35619
CCC(C(=O)O)N
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H317
May cause an allergic skin reaction.

GHS P Statement
P261-P280-P363-P321-P333+P313-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Wash contaminated clothing before reuse.
Specific treatment (see label).
If skin irritation or rash occurs: Get medical advice/attention.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

EINECSNumber : 216-083-3

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only