accessibility menu, dialog, popup

UserName

CH3NH2OHO

CAS RN 1492-24-6

IUPAC Name: 2-aminobutanoic acid
Synonyms: Abu butyrine AABA
Molecular Weight (g/mol): 103.12
Molecular Formula: C4H9NO2
InChi Key: QWCKQJZIFLGMSD-UHFFFAOYNA-N
SMILES: CCC(N)C(O)=O
Molecular Weight (g/mol) 
  • (2)
  • (5)
Percent Purity 
  • (3)
  • (2)
  • (2)
Quantity 
  • (3)
  • (1)
  • (3)
Physical Form 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (5)
  • (2)
  • (5)
  • (3)
  • (2)
  • (2)
  • (3)
  • (1)
  • (3)
  • (4)