missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-(+)-2-Aminobutyric acid, 98+%
Description
L-(+)-2-Aminobutyric acid is used in the biosynthesis of nonribosomal peptides. It acts as a receptor antagonist. It is also used as a chiral reagent. Further, it is used in the determination of substrate of glutamyl cysteine acid synthase. In addition to this, it is also utilized as a drug intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
L-(+)-2-Aminobutyric acid is used in the biosynthesis of nonribosomal peptides. It acts as a receptor antagonist. It is also used as a chiral reagent. Further, it is used in the determination of substrate of glutamyl cysteine acid synthase. In addition to this, it is also utilized as a drug intermediate.
Solubility
Soluble in water and acetic acid. Slightly soluble in alcohol and ether.
Notes
Incompatible with strong oxidizing agents.
L-(+)-2-Aminobutyric acid is used in the biosynthesis of nonribosomal peptides. It acts as a receptor antagonist. It is also used as a chiral reagent. Further, it is used in the determination of substrate of glutamyl cysteine acid synthase. In addition to this, it is also utilized as a drug intermediate.
Solubility
Soluble in water and acetic acid. Slightly soluble in alcohol and ether.
Notes
Incompatible with strong oxidizing agents.
Specifications
Specifications
| Melting Point | >300°C |
| Quantity | 1 g |
| Beilstein | 1720935 |
| Merck Index | 14,428 |
| Solubility Information | Soluble in water and acetic acid. Slightly soluble in alcohol and ether. |
| Optical Rotation | +21° (c=4.8 in 6N HCl) |
| Formula Weight | 103.12 |
| Percent Purity | ≥98% |
| Chemical Name or Material | L-(+)-2-Aminobutyric acid |
Safety and Handling
| Hazard Category | Warning |
| Hazard Statement | GHS H Statement H317 May cause an allergic skin reaction. |
| Precautionary Statement | GHS P Statement P261-P280-P363-P321-P333+P313-P501a Avoid breathing dust/fume/gas/mist/vapors/spray. Wear protective gloves/protective clothing/eye protection/face protection. Wash contaminated clothing before reuse. Specific treatment (see label). If skin irritation or rash occurs: Get medical advice/attention. Dispose of contents/container in accordance with local/regional/national/international regulations. |
| EINECSNumber | 216-083-3 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only