missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Supelco™ Kromasil C18 HPLC Column, 100 Å, 5 μm Particle Size

Catalog No. 111016068
Change view
Click to view available options
Length (Metric):
150 mm
250 mm
Diameter (Metric):
4.6 mm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric) Diameter (Metric)
111016068 150 mm 4.6 mm
111020737 250 mm 4.6 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111016068 Supplier MilliporeSigma™ Supelco™ Supplier No. K08670356

Kromasil C18 HPLC Column, 100 Å, 5 μm Particle Size for small molecules and peptides.

Kromasil Classic 100 Å family of products is used to separate and purify molecules of up to about 10,000 Da.

  • Kromasil™ C18 HPLC column involves a silica-based packing.
  • The reduced levels of Na, K, Ca, Fe, Al, and Mg make the column useful for polar compounds, which tend to interact with the silica surface.
  • Reverse phase (RP) HPLC columns are advantageous in ensuring high precision data, they are widely available commercially and are easy to use and less time-consuming.
  • Two main approaches one should consider before choosing a RP column are, firstly characterising physiochemical parameters of the packing material such as 1% carbon load, surface area, pore size, particle size and pH.
    • These are some of the important parameters important to control the quality of the column.
  • Second approach is based upon the chromatographic and statistical evaluation of the retention behaviour for a large number of compounds.
  • Kromasil C18 columns are basically RP columns made up of silica-based monomeric type reversed-phase packing material.

Specifications

Column Format HPLC Column
Diameter (Metric) 4.6 mm
Length (Metric) 150 mm
Particle Size 5 μm
Pore Size 100 Å
USP Type USP L1
Stationary Phase C18 (octadecyl)
Product Line Kromasil
Matrix C18 (octadecyl) Phase
Packing Material Silica-based Monomeric
Column Type Reversed Phase
Phase Reversed Phase
Technique HPLC
Product Type HPLC Column
Show More Show Less

Kromasil is a registered trademark of EKA Chemicals AB