Learn More
DWK Life Sciences Kimble™ Kontes™ Septum PTFE-Faced Silicone Caps and Closures
Used with open-top cap to access container with a syringe
$213.18 - $240.14
| Catalog Number | Mfr. No. | ForUseWithEquipment | NeckFinish | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | ForUseWithEquipment | NeckFinish | Price | Quantity & Availability | ||||
|
K7741610020
|
DWK Life Sciences
7741610020 |
Case of 24 Each for $213.18
|
|
||||||
|
22311997
|
DWK Life Sciences
7741610024 |
Case of 24 Each for $240.14
|
|
||||||
Description
- PTFE facing maintains the integrity of the contents
- Silicone-rubber backing allows repeated puncturing through the seal
- All septa are 0.165cm (0.065 in.) thick
Specifications
| PTFE-faced Silicone Rubber |