Learn More
DWK Life Sciences Kimble™ Kontes™ Septum PTFE-Faced Silicone Caps and Closures
Used with open-top cap to access container with a syringe
Supplier:  DWK Life Sciences 7741610024
 View more versions of this product
Catalog No. 22311997
Only null left
Description
Description
  • PTFE facing maintains the integrity of the contents
  • Silicone-rubber backing allows repeated puncturing through the seal
  • All septa are 0.165cm (0.065 in.) thick
Specifications
Chemical Identifiers
PTFE-faced Silicone Rubber
Specifications
PTFE-faced Silicone Rubber
24/Cs.
24 mm
Product Suggestions
Product Suggestions