Learn More
DWK Life Sciences™ Kimble™ Kontes™ Distilling Vigreux Column with Standard Taper Joints
Vigreux distilling column with indentations over the entire length for improved vapor-liquid contact. A range of column lengths (130mm, 150mm or 300mm) for a variety of joint sizes.
Supplier:  DWK Life Sciences™ 2867100300
 View more versions of this product
Catalog No. K2867100300
Only null left
Description
Description
  • KIMAX™ column has a Standard Taper outer joint at the top and a Standard Taper inner joint at the bottom
  • Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
  • Options:
    • Column length 130mm, 14/20 Standard Taper joints
    • Column Length 150mm, 24/40 Standard Taper joints
    • Column length 300mm, 19/22 Standard Taper joints
    • Column Length 300mm, 24/40 Standard Taper joints
Specifications
Chemical Identifiers
Vigreux Column
USP Type I, ASTM E438, Type I, Class A
Specifications
300mm
11.8 in.
19/22
Vigreux Column
USP Type I, ASTM E438, Type I, Class A
Product Suggestions
Product Suggestions