Learn More
DWK Life Sciences Kimble™ Kontes™ Brand Rotary Evaporator Glass Vial Connecting Adapters
Ideal for use with small samples
$132.86 - $175.61
Specifications
Material Glass
Product Type Vial Connecting Adapter
View More Specs
Products 3
Catalog Number Mfr. No. Closure Size For Use With (Application) Price Quantity & Availability  
Catalog Number Mfr. No. Closure Size For Use With (Application) Price Quantity & Availability  
K7471301319
View Documents Promotion Details
DWK Life Sciences
7471301319
13-425 Standard Taper Joint: 19/22
Each for $132.86
Only null left
K7471301324
View Documents Promotion Details
DWK Life Sciences
7471301324
13-425 Standard Taper Joint: 24/40
Each for $175.61
Only null left
K7471302024
View Documents Promotion Details
DWK Life Sciences
7471302024
20-400 Standard Taper Joint: 24/40
Each for $174.25
Only null left
Description
Description
Ideal for use with small samples
Connect standard rotary evaporator traps to vials with variety of GPI thread sizes. Typical receivers include Accuform™ 1 to 10mL vials (06-100C, 06-100E, 06-100F, 06-100G). Any standard vial with GPI threads can be used.
Specifications
Specifications
Glass
Vial Connecting Adapter