Learn More
DWK Life Sciences Kimble™ Kontes™ Brand Rotary Evaporator Glass Vial Connecting Adapters
Catalog No. K7471301319
Change view
Click to view available options
Closure Size:
13-425
20-400
For Use With (Application):
Standard Taper Joint: 19/22
Standard Taper Joint: 24/40
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Closure Size For Use With (Application)
K7471301319 13-425 Standard Taper Joint: 19/22
K7471301324 13-425 Standard Taper Joint: 24/40
K7471302024 20-400 Standard Taper Joint: 24/40
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. K7471301319 Supplier DWK Life Sciences Supplier No. 7471301319
Ideal for use with small samples
Connect standard rotary evaporator traps to vials with variety of GPI thread sizes. Typical receivers include Accuform™ 1 to 10mL vials (06-100C, 06-100E, 06-100F, 06-100G). Any standard vial with GPI threads can be used.
Specifications
Material Glass
For Use With (Application) Standard Taper Joint: 19/22
Closure Size 13-425
Product Type Vial Connecting Adapter