Learn More
DWK Life Sciences Kimble™ Kontes™ Brand Rotary Evaporator Glass Vial Connecting Adapters
Catalog No. K7471302024
Change view
Click to view available options
Closure Size:
13-425
20-400
For Use With (Application):
Standard Taper Joint: 24/40
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Closure Size For Use With (Application)
K7471302024 20-400 Standard Taper Joint: 24/40
K7471301324 13-425 Standard Taper Joint: 24/40
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. K7471302024 Supplier DWK Life Sciences Supplier No. 7471302024
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Ideal for use with small samples
Connect standard rotary evaporator traps to vials with variety of GPI thread sizes. Typical receivers include Accuform™ 1 to 10mL vials (06-100C, 06-100E, 06-100F, 06-100G). Any standard vial with GPI threads can be used.
Specifications
Material Glass
For Use With (Application) Standard Taper Joint: 24/40
Closure Size 20-400
Product Type Vial Connecting Adapter