Learn More
DWK Life Sciences Kimble™ Kontes™ Brand 5mm High Grade NMR Sample Tubes
Each tube meets rigid specifications for wall thickness, variation and camber so as to assure precise measurements and reproducibility.
$78.06 - $1,072.39
Specifications
| Diameter (English) Outer | 0.19 in. |
|---|---|
| Material | Borosilicate Glass |
| Closure Material | Polyethylene |
| Closure Type | Compression-fit |
| Diameter (Metric) Outer | 5 mm |
| missing translation for 'productCode' | missing translation for 'brand' | Length (English) | Field Strength | Camber (English) | Concentricity (English) | Price | Quantity & Availability | ||
|---|---|---|---|---|---|---|---|---|---|
| missing translation for 'productCode' | missing translation for 'brand' | Length (English) | Field Strength | Camber (English) | Concentricity (English) | Price | Quantity & Availability | ||
|
K8972200000
|
DWK Life Sciences
8972200000 |
7 in. | 150MHz | ±0.001 in. | ±0.002 in. |
Case of 5 Each for $111.66
|
|
||
|
K8972000000
|
DWK Life Sciences
8972000000 |
7 in. | 100MHz | ±0.002 in. | ±0.003 in. |
Pack of 5 Each for $78.06
|
|
||
|
K8972000008
|
DWK Life Sciences
8972000008 |
8 in. | 100MHz | ±0.002 in. | ±0.003 in. |
Case of 5 Each for $107.61
|
|
||
|
K8972050000
|
DWK Life Sciences
8972050000 |
7 in. | 100MHz | ±0.002 in. | ±0.0025 in. |
Pack of 5 Each for $163.50
|
|
||
|
K8972200008
|
DWK Life Sciences
8972200008 |
8 in. | 150MHz | ±0.0011 in. | ±0.002 in. |
Case of 5 Each for $122.90
|
|
||
|
K8972050008
|
DWK Life Sciences
8972050008 |
8 in. | 100MHz | ±0.002 in. | ±0.0025 in. |
Case of 5 Each for $102.78
|
|
||
|
K8972250008
|
DWK Life Sciences
8972250008 |
8 in. | 200MHz | ±0.0006 in. | ±0.002 in. |
Case of 5 Each for $163.22
|
|
||
|
K8972300000
|
DWK Life Sciences
8972300000 |
7 in. | 300MHz | ±0.001 in. | ±0.0015 in. |
Pack of 5 Each for $147.06
|
|
||
|
K8972300008
|
DWK Life Sciences
8972300008 |
8 in. | 300MHz | ±0.0011 in. | ±0.0015 in. |
Pack of 5 Each for $163.22
|
|
||
|
K8972350008
|
DWK Life Sciences
8972350008 |
8 in. | 400MHz | ±0.0006 in. | ±0.001 in. |
Pack of 5 Each for $312.36
|
|
||
|
K8972400009
|
DWK Life Sciences
8972400009 |
9 in. | 500MHz | ±0.00045 in. | ±0.0005 in. |
Case of 5 Each for $260.44
|
|
||
|
K8972350009
|
DWK Life Sciences
8972350009 |
9 in. | 400MHz | ±0.0007 in. | ±0.001 in. |
Pack of 5 Each for $194.72
|
|
||
|
K8972050009
|
DWK Life Sciences
8972050009 |
9 in. | 100MHz | ±0.002 in. | ±0.0025 in. |
Case of 5 Each for $116.34
|
|
||
|
K8972250009
|
DWK Life Sciences
8972250009 |
9 in. | 200MHz | ±0.0007 in. | ±0.002 in. |
Pack of 5 Each for $186.20
|
|
||
|
K8972503000
|
DWK Life Sciences
8972503000 |
7 in. | 900MHz | ±0.00015 in. | ±0.0002 in. |
Pack of 5 Each for $1,072.39
|
|
||
|
K8972410009
|
DWK Life Sciences
8972410009 |
9 in. | 600 to 700MHz | ±0.00045 in. | ±0.0005 in. |
Case of 5 Each for $626.61
|
|
||
|
K8972250000
|
DWK Life Sciences
8972250000 |
7 in. | 200MHz | ±0.0005 in. | ±0.002 in. |
Case of 5 Each for $147.06
|
|
||
|
K8972350000
|
DWK Life Sciences
8972350000 |
7 in. | 400MHz | ±0.0005 in. | ±0.001 in. |
Case of 5 Each for $172.88
|
|
||
|
K8972400000
|
DWK Life Sciences
8972400000 |
7 in. | 500MHz | ±0.00025 in. | ±0.0005 in. |
Case of 5 Each for $216.62
|
|
||
|
K8972400008
|
DWK Life Sciences
8972400008 |
8 in. | 500MHz | ±0.00035 in. | ±0.0005 in. |
Case of 5 Each for $423.61
|
|
||
Description
Precision-manufactured tubes designed for use in high-resolution NMR spectroscopy that characterizes organic molecules through measurement of the resonant frequencies of atomic nuclei. Research applications include analysis of polymer structures and organic solutions.
- Made from Type I borosilicate glass
- Wall thickness: 0.01475 in. (0.37mm)
- Wall variation and camber depend, in part, on tube length
- All sample tubes come with plastic compression-fit caps and sandblasted marking spots
Specifications
| 0.19 in. | |
| Polyethylene | |
| 5 mm | |
| Ideal for sealing directly to vacuum manifolds, joints or valves | |
| 0.014 in. |
| Borosilicate Glass | |
| Compression-fit | |
| 4.2 mm | |
| Cap | |
| 0.38 mm |