missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Supelco™ Knitted Reactor Coil

Catalog No. 111004251
Change view
Click to view available options
Volume (Metric):
0.25 mL
0.5 mL
0.75 mL
1 mL
2 mL
3 mL
4 mL
Description:
L I.D. 10 m 0.25 mm
L I.D. 10 m 0.5 mm
L I.D. 15 m 0.25 mm
L I.D. 15 m 0.5 mm
L I.D. 20 m 0.25 mm
L I.D. 20 m 0.5 mm
L I.D. 5 m 0.25 mm
L I.D. 5 m 0.5 mm
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Description
111004251 0.25 mL L I.D. 5 m 0.25 mm
111003827 0.5 mL L I.D. 10 m 0.25 mm
111003672 0.75 mL L I.D. 15 m 0.25 mm
111016300 1 mL L I.D. 5 m 0.5 mm
111016184 1 mL L I.D. 20 m 0.25 mm
111003749 2 mL L I.D. 10 m 0.5 mm
111003496 3 mL L I.D. 15 m 0.5 mm
111003500 4 mL L I.D. 20 m 0.5 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 111004251 Supplier MilliporeSigma™ Supelco™ Supplier No. SGMA57402

For use with PHRED Reactor

Specifications

Description L I.D. 5 m 0.25 mm
For Use With (Equipment) PHRED Reactor
Volume (Metric) 0.25 mL