missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ KN 92

Catalog No. 41301
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
41301 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 41301 Supplier Tocris Bioscience™ Supplier No. 4130/1

Inactive analog of KN 93. K+ channel (Kv) blocker

Inactive analog of the CaM kinase II inhibitor KN 93 . Potassium channel (Kv1.2, 1.4, 1.5, 2.1, 3.2 and hERG) blocker in vitro.

Active Analog also available.

Chemical Identifiers

CAS 1135280-28-2
Molecular Formula C24H28ClN2O7PS

Specifications

Quantity 1 mg
Formula Weight 554.98
Molecular Weight (g/mol) 554.979
Chemical Name or Material KN 92
Recommended Storage Store at -20°C