missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ KJ Pyr 9

Catalog No. 530650
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
530650 50 mg
530610 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 530650 Supplier Tocris Bioscience™ Supplier No. 5306/50
Only null left

High affinity Myc inhibitor

KJ Pyr 9 is a high affinity Myc inhibitor (Kd = 6.5 nM). Disrupts Myc-Max interaction. Inhibits Myc-dependent cancer cell line proliferation in vitro. Inhibits growth of MDA-MB-231 breast cancer cell xenografts in mice. Brain penetrant.

Specifications

Chemical Name or Material 4-[2-(2-Furanyl)-6-(4-nitrophenyl)-4-pyridinyl]benzamide
CAS 581073-80-5
Quantity 50 mg
Target Additional Transcription Factor Compounds
Molecular Formula C22H15N3O4
Formula Weight 385.37
Purity >98%