missing translation for 'onlineSavingsMsg'
Learn More

Kininogen-1 Armenian Hamster anti-Human, Clone: 300000000, MilliporeSigma™

Armenian Hamster Monoclonal Antibody

$298.74 - $756.86

Specifications

Antigen Kininogen-1
Clone 300000000
Applications ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation
Classification Monoclonal
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
MABN260825
View Documents
MilliporeSigma
MABN260825UG
25 μg
Each for $298.74
Only null left
 
MABN2608100
View Documents
MilliporeSigma
MABN2608100UG
100 μg
Each for $756.86
Only null left
 
Description

Description

Kininogen-1 Monoclonal antibody specifically detects Kininogen-1 in Human samples. It is validated for ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation.
Specifications

Specifications

Kininogen-1
ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation
Unconjugated
Armenian Hamster
Human
KLH-conjugated linear peptide corresponding to 19 amino acids from the C-terminal region of human Kininogen-1.
Protein G purified
300000000
Monoclonal
Purified
RUO
Purified armenian Hamster monoclonal antibody IgG2-IgG3 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Primary
Stable for 1 year at 2-8°C from date of receipt.
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'