missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kininogen-1 Armenian Hamster anti-Human, Clone: 300000000, MilliporeSigma™
Armenian Hamster Monoclonal Antibody
Supplier: MilliporeSigma MABN260825UG
Description
Kininogen-1 Monoclonal antibody specifically detects Kininogen-1 in Human samples. It is validated for ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation.Specifications
| Kininogen-1 | |
| Monoclonal | |
| Unconjugated | |
| Armenian Hamster | |
| Protein G purified | |
| RUO | |
| Human | |
| Purified |
| ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation | |
| 300000000 | |
| Purified armenian Hamster monoclonal antibody IgG2-IgG3 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide. | |
| KLH-conjugated linear peptide corresponding to 19 amino acids from the C-terminal region of human Kininogen-1. | |
| 25 μg | |
| Primary | |
| Stable for 1 year at 2-8°C from date of receipt. |