missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ KingFisher™ Flex™ Purification System Accessories

Prepare samples efficiently from a variety of materials with Thermo Scientific™ KingFisher™ Flex Purification Systems which offer highly versatile, automated magnetic-particle processing for DNA/RNA, protein, or cell purification from virtually any source.

$990.00 - $11,733.87

Specifications

For Use With (Equipment) KingFisher™ Flex™ Purification System
View More Specs

Products 7
Catalog Number Mfr. No. Product Type Price Quantity & Availability  
Catalog Number Mfr. No. Product Type Price Quantity & Availability  
22387009
Encompass
View Documents Promotion Details
Thermo Scientific™
24074411
Flex 96 PCR Head, heat block N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
22387017
Encompass
View Documents Promotion Details
Thermo Scientific™
24075410
Flex 96 PCR heating block
Each for $990.00
Only null left
 
22387014
Encompass
View Documents Promotion Details
Thermo Scientific™
24074420
Flex KF Head
Each for $8,780.00
Only null left
 
22387013
Encompass
View Documents Promotion Details
Thermo Scientific™
24074410
Flex 96 PCR Head
Each for $8,600.00
Only null left
 
22387012
Encompass
View Documents Promotion Details
Thermo Scientific™
24074441
Flex 24 DW head, heating block
Each for $9,150.00
Only null left
 
22387010
Encompass
View Documents Promotion Details
Thermo Scientific™
24074421
Flex 96 KF Head, heating block
Each for $9,260.00
Only null left
 
22387011
Encompass
View Documents Promotion Details
Thermo Scientific™
24074431
Flex 96 DW Head, heating block N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Find a flexible solution for different types of sample processing needs. With 24 magnet heads, customers can raise the processing volume up to 5 mL, resulting in increased yield of the purified product. For higher throughput needs, 96 samples can be processed with 96 magnet head.

  • Fully automated system yields high-speed purification of nucleic acids, proteins and cells.
  • High throughput system processes up to 96 samples in less than 20 minutes.
  • Open and flexible system lets the customer use any magnetic particle-based kit to meet the application demands.
  • Easy-to-use Thermo Scientific™ BindIt™ Software provides instrument control, protocol creation and modification.
  • Ready-made protocols for different type of applications are available.

Recommended for:

  • Genomics and proteomics
  • Target identification
  • Veterinary assays
  • Biomarker discovery
  • Quality control
  • Forensics
Specifications

Specifications

KingFisher™ Flex™ Purification System
Videos
SDS
Documents

Documents