missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ KingFisher™ Flex™ Purification System Accessories
Description
Find a flexible solution for different types of sample processing needs. With 24 magnet heads, customers can raise the processing volume up to 5 mL, resulting in increased yield of the purified product. For higher throughput needs, 96 samples can be processed with 96 magnet head.
- Fully automated system yields high-speed purification of nucleic acids, proteins and cells.
- High throughput system processes up to 96 samples in less than 20 minutes.
- Open and flexible system lets the customer use any magnetic particle-based kit to meet the application demands.
- Easy-to-use Thermo Scientific™ BindIt™ Software provides instrument control, protocol creation and modification.
- Ready-made protocols for different type of applications are available.
Recommended for:
- Genomics and proteomics
- Target identification
- Veterinary assays
- Biomarker discovery
- Quality control
- Forensics
Specifications
Specifications
| Product Type | Flex 96 PCR Head |
| For Use With (Equipment) | KingFisher™ Flex™ Purification System |