missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Kinesin 5B Recombinant Rabbit Monoclonal Antibody (23GB4030)

Rabbit Recombinant Monoclonal Antibody
Specifications
| Antigen | Kinesin 5B |
|---|---|
| Clone | 23GB4030 |
| Concentration | 0.27 mg/mL |
| Content And Storage | -20°C |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
Description
Kinesin 5B Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
Microtubule-dependent motor required for normal distribution of mitochondria and lysosomes. Can induce formation of neurite-like membrane protrusions in non-neuronal cells in a ZFYVE27-dependent manner. Regulates centrosome and nuclear positioning during mitotic entry. During the G2 phase of the cell cycle in a BICD2-dependent manner, antagonizes dynein function and drives the separation of nuclei and centrosomes. Required for anterograde axonal transportation of MAPK8IP3/JIP3 which is essential for MAPK8IP3/JIP3 function in axon elongation.Specifications
| Kinesin 5B | |
| 0.27 mg/mL | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P33176, Q2PQA9, Q61768 | |
| 117550, 16573, 3799 | |
| A synthesized peptide derived from human KIF5B (10-45AA). | |
| Antibody |
| 23GB4030 | |
| -20°C | |
| Recombinant Monoclonal | |
| Liquid | |
| IgG | |
| Human, Mouse, Rat | |
| KIF5B | |
| AL022807; Conventional kinesin heavy chain; EMBL:ABC25059.1}; epididymis secretory protein Li 61; HEL-S-61; Khc; Khcs; KIF5A; KIF5B; kif5b {ECO:0000312; kinesin 1 (110-120kD); kinesin family member 5A; kinesin family member 5B; kinesin heavy chain; kinesin heavy chain member 5B, ubiquitous; kinesin-1 heavy chain; KINH; KNS; Kns1; ubiquitous kinesin heavy chain; UKHC; U-KHC; Unknown (protein for IMAGE:7944745) | |
| KIF5B | |
| Primary | |
| Affinity chromatography |