missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Kinesin 5B Recombinant Rabbit Monoclonal Antibody (23GB4030)

Catalog No. MA553675
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA553675 20 μL
MA553674 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA553675 Supplier Invitrogen™ Supplier No. MA553675

Rabbit Recombinant Monoclonal Antibody

Kinesin 5B Recombinant Monoclonal Antibody for Flow, ICC/IF, Western Blot

Microtubule-dependent motor required for normal distribution of mitochondria and lysosomes. Can induce formation of neurite-like membrane protrusions in non-neuronal cells in a ZFYVE27-dependent manner. Regulates centrosome and nuclear positioning during mitotic entry. During the G2 phase of the cell cycle in a BICD2-dependent manner, antagonizes dynein function and drives the separation of nuclei and centrosomes. Required for anterograde axonal transportation of MAPK8IP3/JIP3 which is essential for MAPK8IP3/JIP3 function in axon elongation.

Specifications

Antigen Kinesin 5B
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 23GB4030
Concentration 0.27 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene KIF5B
Gene Accession No. P33176, Q2PQA9, Q61768
Gene Alias AL022807; Conventional kinesin heavy chain; EMBL:ABC25059.1}; epididymis secretory protein Li 61; HEL-S-61; Khc; Khcs; KIF5A; KIF5B; kif5b {ECO:0000312; kinesin 1 (110-120kD); kinesin family member 5A; kinesin family member 5B; kinesin heavy chain; kinesin heavy chain member 5B, ubiquitous; kinesin-1 heavy chain; KINH; KNS; Kns1; ubiquitous kinesin heavy chain; UKHC; U-KHC; Unknown (protein for IMAGE:7944745)
Gene Symbols KIF5B
Host Species Rabbit
Immunogen A synthesized peptide derived from human KIF5B (10-45AA).
Purification Method Affinity chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 117550, 16573, 3799
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less