missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Kimble™ Phenolic Caps with Solid PE Liners

Catalog No. 13757137
Change view
Click to view available options
Neck Finish:
20-400
22-400
24-400
28-400
33-400
38-400
43-400
45-400
48-400
53-400
58-400
63-400
70-400
89-400
14 product options available for selection
Product selection table with 14 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Neck Finish
13757137 58-400
13757140 89-400
13757139 70-400
13757130 28-400
13757133 43-400
13757128 22-400
13757129 24-400
13757132 38-400
13757135 48-400
13757131 33-400
13757138 63-400
13757134 45-400
13757136 53-400
13757127 20-400
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 14 options available.
14 options
Not recommended for hydrocarbon solvents.
Catalog No. 13757137 Supplier DWK Life Sciences Supplier No. 75202G58400
Not recommended for hydrocarbon solvents.

Has good resistance to acids, alcohols, alkalis, aqueous solutions, oils, solvents

  • General purpose phenolic screw cap with a solid polyethylene (PE) liner
  • Color: black
  • Liner material: solid polyethylene (0.040-in.)
  • Ideal for use with strong acid or base samples
  • Not recommended for hydrocarbon solvents

Specifications

Color Black
Material Phenolic Resin
No. per Case 144 Cs.
Neck Finish 58-400
Product Type Phenolic Cap
For Use With (Equipment) Bottle
Closure Liner Polyethylene