missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences Kimble™ Phenolic Caps with 14B White Rubber Liners
Resilient and resistant to moisture vapor; provides a chemical barrier
$211.85 - $1,224.54
Specifications
| Color | Black |
|---|---|
| Material | Phenolic Resin |
| No. per Case | 144 Cs. |
| Product Type | Phenolic Cap |
| For Use With (Equipment) | Bottle |
| Catalog Number | Mfr. No. | Neck Finish | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Neck Finish | Price | Quantity & Availability | |||||
|
13757156
|
DWK Life Sciences
75204G22400 |
22-400 |
Case of 144 Each for $211.85
|
|
|||||
|
13757155
|
DWK Life Sciences
75204G20400 |
20-400 |
Case of 144 Each for $260.96
|
|
|||||
|
13757158
|
DWK Life Sciences
75204G28400 |
28-400 |
Case of 144 Each for $282.32
|
|
|||||
|
13757157
|
DWK Life Sciences
75204G24400 |
24-400 |
Case of 144 Each for $309.32
|
|
|||||
|
13757160
|
DWK Life Sciences
75204G38400 |
38-400 |
Case of 144 Each for $295.92
|
|
|||||
|
13757161
|
DWK Life Sciences
75204G43400 |
43-400 |
Case of 144 Each for $460.25
|
|
|||||
|
13757162
|
DWK Life Sciences
75204G45400 |
45-400 |
Case of 144 Each for $412.92
|
|
|||||
|
13757163
|
DWK Life Sciences
75204G48400 |
48-400 |
Case of 144 Each for $449.92
|
|
|||||
|
13757164
|
DWK Life Sciences
75204G53400 |
53-400 |
Case of 144 Each for $727.43
|
|
|||||
|
13757168
|
DWK Life Sciences
75204G89400 |
89-400 |
Case of 144 Each for $1,224.54
|
|
|||||
|
13757165
|
DWK Life Sciences
75204G58400 |
58-400 |
Case of 144 Each for $600.41
|
|
|||||
|
13757166
|
DWK Life Sciences
75204G63400 |
63-400 |
Case of 144 Each for $741.46
|
|
|||||
Description
- Resilient
- Resistant to moisture vapor
- Hermetic sealing for biologicals and other contents requiring sterilization
- 14B white rubber provides chemical barrier
All caps/liners should be tested to ensure leak and compatibility performance with contents.
Specifications
| Black | |
| 144 Cs. | |
| Bottle |
| Phenolic Resin | |
| Phenolic Cap | |
| PTFE/14B Rubber |