Learn More
DWK Life Sciences Kimble™ Phenolic Caps with 14B White Rubber Liners
Resilient and resistant to moisture vapor; provides a chemical barrier
Supplier: DWK Life Sciences 75204G43400
Description
- Liner material: homogenous white rubber (0.050-inch)
- Use for hermetic sealing for biologicals and other contents requiring sterilization
- Autoclavable
Chemical Identifiers
| Black | |
| 144 Cs. | |
| Bottle |
| Phenolic Resin | |
| Phenolic Cap | |
| PTFE/14B Rubber |
Specifications
| Black | |
| 144 Cs. | |
| Phenolic Cap | |
| PTFE/14B Rubber |
| Phenolic Resin | |
| 43-400 | |
| Bottle |