missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Kimble™ Phenolic Caps with 14B White Rubber Liners

Catalog No. 13757156
Change view
Click to view available options
Neck Finish:
20-400
22-400
24-400
28-400
38-400
43-400
45-400
48-400
53-400
58-400
63-400
89-400
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Neck Finish
13757156 22-400
13757155 20-400
13757158 28-400
13757157 24-400
13757160 38-400
13757161 43-400
13757162 45-400
13757163 48-400
13757164 53-400
13757168 89-400
13757165 58-400
13757166 63-400
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
Catalog No. 13757156 Supplier DWK Life Sciences Supplier No. 75204G22400

Resilient and resistant to moisture vapor; provides a chemical barrier

  • Liner material: homogenous white rubber (0.050-inch)
  • Use for hermetic sealing for biologicals and other contents requiring sterilization
  • Autoclavable

Specifications

Color Black
Material Phenolic Resin
No. per Case 144 Cs.
Neck Finish 22-400
Product Type Phenolic Cap
For Use With (Equipment) Bottle
Closure Liner PTFE/14B Rubber