Learn More
DWK Life Sciences Kimble™ Kontes™ Open-Top Compression Caps for Microscale Chemistry Kits
For use with MICROFLEX™ microscale chemistry kits
Supplier:  DWK Life Sciences 4101192015
 View more versions of this product
Catalog No. K410119-2015
Only null left
Explore available promotions
Description
Description
  • Glass-filled nylon cap with modified GPI thread finish
  • Accommodate us with an O-ring seal
  • Temperature limit: 150°C
Specifications
Specifications
MICROFLEX™ microscale chemistry kits, for 14/10 connections
Nylon
20-400
Closure
Product Suggestions
Product Suggestions